Anti-FOXP2 Recombinant Antibody [Clone: RAB-S249]
Catalog # PRSI24-865
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:forkhead box P2
- Antigen symbol:FOXP2
- Clonality:Recombinant
- Clone:RAB-S249
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:Fab fragment kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:93986
- Antigen synonyms:Trinucleotide repeat-containing gene 10 protein|CAG repeat protein 44|anti-FOXP2-RAB-S249|S0014_B01_HR7934B.006_A10|Forkhead box protein P2
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP2 protein under non-denaturing conditions
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: forkhead box P2
Clonality: Recombinant
Clone: RAB-S249
Conjugation: Unconjugated
Host: Recombinant
Isotype: Fab fragment kappa
Reactivity: Human