Anti-CD37 Recombinant Antibody [Clone: IPO-24 (ZC37-24)]
Catalog # PRSI24-753
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD37 molecule
- Antigen symbol:CD37
- Clonality:Recombinant
- Clone:IPO-24 (ZC37-24)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:951
- Antigen synonyms:Tspan-26|Tetraspanin-26|tetraspanin|Gp52-40|Leukocyte antigen CD37
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunizing mice with spleen cells of a patient with hairy cell leukemia
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: CD37 molecule
Clonality: Recombinant
Clone: IPO-24 (ZC37-24)
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human