
Iron(III) sulphate hydrate
Catalog # SIAL307718-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle, wide mouth
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay:97%
Specification Test Results
About this item
:
:399.88 g/mol (anhy)
Merck Index:13,04065
Density:3,1 g/cm³ (20 °C)
EINECS:233-072-9
:GHS07
Storage Temperature:Ambient
CAS Number:15244-10-7
Formula:Fe₂(SO₄)₃·xH₂O
MDL Number:MFCD00149714