


Hexachloroplatinic(IV) acid hexahydrate ACS
Catalog # SIAL206083-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:517.9 g/mol
Merck Index:13,07609
Melting Pt:150 °C
Density:2,434 g/cm³ (20 °C)
EINECS:241-010-7
UN:2507
ADR:8,III
:GHS06
:GHS05
:GHS08
CAS Number:18497-13-7
Formula:H₂PtCl₆·6H₂O
MDL Number:MFCD00149910