Human Trypsin (from pancreas)
Catalog # FITZ30-AT45
Supplier: FITZGERALD INDUSTRIES INTL
CAS Number:
Specifications
- Pk:100 µG
- CAS No.:9002-07-7
- Source:Pancreas
- Species:Human
- Environmentally Preferable:
- Storage conditions:Refrigerator
- MDL No.:MFCD00082094
- Enzyme name:Trypsin