Trypsin
100 µG
Catalog # FITZ70-XR77
Supplier: FITZGERALD INDUSTRIES INTL
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:100 µG
- Cat. No.:FITZ70-XR77
- Storage conditions:Refrigerator
- MDL No.:MFCD00082094
- Enzyme name:Trypsin
- CAS No.:9002-07-7



