Order Entry
Export
ContactUsLinkComponent
 
 
(±)-trans-1,2-Diaminocyclohexane 98%
 
undefined
 
(±)-trans-1,2-Diaminocyclohexane 98%

 

  • Appearance:
    Clear colorless to yellow Liquid (May develop some turbidity or precipitate)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4870 to 1.4900 (20°C, 589 nm)
  • GC:
    ≥97.5 %