
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Catalog # EHERC12194000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:364.35 g/mol
Melting Pt:> 210 °C
EINECS:236-308-9
:GHS07
CAS Number:125572-95-4
:MFCD00066429
Formula:C₁₄H₂₂N₂O₈·1H₂O