Riboflavine (vitamin B2), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:83-88-5
:
Formula:C₁₇H₂₀N₄O₆
MDL Number:MFCD00005022
:376.37 g/mol
Merck Index:12,08367
Melting Pt:280 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:201-507-1