Order Entry
Export
ContactUsLinkComponent
Phenol red sodium salt
Phenol red sodium salt
Catalog # MOLE45234408-25G
Supplier:  Molekula
CAS Number:  
Phenol red sodium salt
Catalog # MOLE45234408-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :376.35 g/mol
Merck Index:12,07397
Melting Pt:285 °C
Storage Temperature:Ambient
EINECS:252-057-8
CAS Number:34487-61-1
Formula:C₁₉H₁₃NaO₅S
MDL Number:MFCD00066901