Order Entry
Export
Orders LinkContactUsLinkComponent
Methanol

Methanol

Glass bottle, 1 L
Catalog # ROTHHN41.1
Flammable
Toxic
Health
   Safer Human and Environmental Health
Supplier:   Roth Carl
Methanol
Catalog # ROTHHN41.1
Supplier:  Roth Carl
   Safer Human and Environmental Health
CAS Number:  67-56-1
Methanol
Methanol

Glass bottle, 1 L

CAS Number:  67-56-1
Price
46,60 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64.6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Cat. No.:
    ROTHHN41.1
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications