

Erbium(III) perchlorate 40% (w/w) in aqueous solution ≥99.9% (trace metals basis), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:14017-55-1
:
Formula:Cl₃ErO₁₂
MDL Number:MFCD00016073
:465.61 g/mol
ADR:5.1,II
:M-Clarity™ quality level = MQ100
EINECS:237-840-4
UN:3211
:GHS03
:GHS07