Sodium alginate
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Alginic acid sodium salt, Algin
- CAS Number:9005-38-3
- MDL Number:MFCD00081310
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: Alginic acid sodium salt, Algin
- CAS Number:9005-38-3
- MDL Number:MFCD00081310
- Storage Temperature:Ambient



