Sodium alginate
Glass bottle for solids, 1 kg
Catalog # MOLE60689063-1KG
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Alginic acid sodium salt, Algin
- CAS Number:9005-38-3
- MDL Number:MFCD00081310
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



