Rhodamine B
Glass bottle for solids, 100 g
Catalog # MOLE16917624
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
- CAS Number:81-88-9
- Formula:C₂₈H₃₁ClN₂O₃
- MDL Number:MFCD00011931
- Molecular Weight:479.02 g/mol
- Merck Index:13,08266
- Melting Pt:199…201 °C
- Density:1.31 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:201-383-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



