Order Entry
Export
Orders LinkContactUsLinkComponent
Rhodamine B

Rhodamine B

Corrosive
Irritant
Rhodamine B
Certificates
Rhodamine B

3 Options Available Below

Product Details & Documents

CAS No.: 81-88-9 Molecular Formula: C28H31ClN2O3 Formula Weight: 479.02 Physical Form: Crystals Density: 1.31 Appearance: Purple powder/dark green Melting Point: ca 210° dec. Beilstein Reference: 4119648 Merck Reference: 14,8183 MDL No.: MFCD00011931

Suitable for fluorescent labelling

Synonyms:  9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
  • CAS Number:81-88-9
  • Formula:C₂₈H₃₁ClN₂O₃
  • MDL Number:MFCD00011931
  • Molecular Weight:479.02 g/mol
  • Merck Index:14,8183
  • Melting Pt:∼ 210 °C (dec.)
  • Density:1.31
  • Storage Temperature:Ambient
  • EINECS:201-383-9
Product Family Options

Product Information