Order Entry
Export
Orders LinkContactUsLinkComponent
Rhodamine B, pure

Rhodamine B, pure

Corrosive
Irritant
Rhodamine B, pure
Certificates
Rhodamine B, pure

3 Options Available Below

Product Details & Documents
Synonyms:  9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
  • CAS Number:81-88-9
  • Formula:C₂₈H₃₁ClN₂O₃
  • MDL Number:MFCD00011931
  • Molecular Weight:479.02 g/mol
  • Merck Index:13,08266
  • Melting Pt:210…211 °C (dec.)
  • Density:1,31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-383-9

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Dark green Powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤10 % (105°C)
  • UV Lambda max:
    549 to 558 nm (in 50:50 IMS:water)
  • Absorption Ratio P-15/P+15:
    1.1 to 1.6
  • Specific absorbance A (1%/1cm):
    ≥1700 (at lambda max.)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 79,00 €
    79,00 €
    1 * 100 g
       79,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 254,00 €
    254,00 €
    1 * 500 g
       254,00 €

  • Plastic drum
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 915,00 €
    915,00 €
    1 * 2,5 kg
       915,00 €