Order Entry
Export
Orders LinkContactUsLinkComponent
Bromophenol blue sodium salt

Bromophenol blue sodium salt

Glass bottle for solids, 10 g
Catalog # ROTHA512.1
Supplier:   Roth Carl
Bromophenol blue sodium salt
Catalog # ROTHA512.1
Supplier:  Roth Carl
CAS Number:  34725-61-6
Bromophenol blue sodium salt

Glass bottle for solids, 10 g

CAS Number:  34725-61-6
Price
58,10 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt
  • CAS Number:34725-61-6
  • Formula:C₁₉H₉Br₄NaO₅S
  • MDL Number:MFCD00013793
  • Molecular Weight:691.95 g/mol
  • Storage Temperature:Ambient
  • EINECS:252-170-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications