Order Entry
Export
ContactUsLinkComponent
Bromocresol green sodium salt 0.5 g/l in ethanol 20% (v/v) Reag. Ph. Eur. 1012601
Bromocresol green sodium salt 0.5 g/l in ethanol 20% (v/v) Reag. Ph. Eur. 1012601
Catalog # 87791.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bromocresol green sodium salt 0.5 g/l in ethanol 20% (v/v) Reag. Ph. Eur. 1012601
Catalog # 87791.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :720 g/mol
CAS Number:62625-32-5
 :67763-24-0
Formula:C₂₁H₁₃Br₄NaO₅S
MDL Number:MFCD00148898