Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bromocresol green sodium salt
Bromocresol green sodium salt, Sigma-Aldrich®
Bromocresol green sodium salt, Sigma-Aldrich®
SIAL17480-25G
:
Merck
:
Bromocresol green sodium salt, Sigma-Aldrich®
SIAL17480-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
225 °C
:
720 g/mol
:
Ambient
:
263-657-4
:
62625-32-5
:
67763-24-0
:
C₂₁H₁₃Br₄NaO₅S
:
MFCD00148898