Order Entry
Export
Orders LinkContactUsLinkComponent
 

Methyl red ACS

Methyl red ACS
Certificates
Methyl red ACS

3 Options Available Below

Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:12,06199
  • Melting Pt:178…182 °C
  • Density:1,31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1
Specifications for Product Family

Specification Test Results

  • Appearance:
    Dark red to brown to purple Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    179 to 182 °C
  • Visual transition interval:
    From pH 4.2 (pink) to pH 6.2 (yellow)
  • Clarity of alcohol solution:
    Passes test
  • Specific optical rotation:
    +52.0° to +62.0° (20 °C, 589 nm) (neat)
Product Family Options

Product Information