Order Entry
Export
Orders LinkContactUsLinkComponent
 

Thiazine red

Glass bottle, 1 L
Catalog # CTEH62062/1
Irritant
Supplier:   CLIN TECH
Thiazine red
Catalog # CTEH62062/1
Supplier:  CLIN TECH
CAS Number:  2150-33-6
Thiazine red

Glass bottle, 1 L

CAS Number:  2150-33-6
Price
51,40 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Disodium 2-[4-[(1-hydroxy-4-sulphonato-2-naphthyl)azo]phenyl]-6-methylbenzothiazole-7-sulphonate
  • CAS Number:2150-33-6
  • Formula:H₂NCOCH₂NHCH₂CH₂SO₃H
  • Molecular Weight:599.58 g/mol
  • Storage Temperature:Ambient
  • EINECS:218-422-0
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications