2,4-Dimethylstyrene, Sigma-Aldrich®
SIAL262633-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:132.21 g/mol
:70 °C (100 mmHg)
:–64.32 °C
:0.905 g/cm³ (20 °C)
:218-781-3
:3295
:3,III
:M-Clarity™ quality level = MQ100
:2234-20-0
:C₁₀H₁₂
:MFCD00014937