Colchicine
Glass bottle for solids, 100 mg
Catalog # PROOMM1083.00
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine
- CAS Number:64-86-8
- Formula:C₂₂H₂₅NO₆
- MDL Number:MFCD00078484
- Molecular Weight:399.44 g/mol
- Merck Index:12,02536
- Melting Pt:154…156 °C
- Density:1.32 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-598-5
- UN:1544
- ADR:6.1,I
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



