Order Entry
Export
ContactUsLinkComponent
Herbicide reference standard, Chlorotoluron (3-(3-Chloro-p-tolyl)-1,1-dimethylurea) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Herbicide reference standard, Chlorotoluron (3-(3-Chloro-p-tolyl)-1,1-dimethylurea) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERXA11530000AL
 :  
Herbicide reference standard, Chlorotoluron (3-(3-Chloro-p-tolyl)-1,1-dimethylurea) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERXA11530000AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    15545-48-9
  • Description:
    Chlorotoluron (3-(3-Chloro-p-tolyl)-1,1-dimethylurea) 100 µg/ml in acetonitrile
  • Formula:
    C₁₀H₁₃ClN₂O
  • Description:
    Chlorotoluron (3-(3-Chloro-p-tolyl)-1,1-dimethylurea)