Order Entry
Export
ContactUsLinkComponent
Chlorotoluron(3-(3-chloro-p-tolyl)-1,1-dimethylurea)
Health
Environment
Chlorotoluron(3-(3-chloro-p-tolyl)-1,1-dimethylurea)
Catalog # EHERC11530000
Supplier:  EHRENSTORFER
CAS Number:  
Health
Environment
Chlorotoluron(3-(3-chloro-p-tolyl)-1,1-dimethylurea)
Catalog # EHERC11530000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :212.68 g/mol
Melting Pt:159…160 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:239-592-2
UN:3077
ADR:9,III
 :GHS08
 :GHS09
CAS Number:15545-48-9
Formula:(C₆H₃ClCH₃)NC(S)NH₂