Monosodium urate, crystals
Catalog # ENZOALX400047M002
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:2 mg
- Seq:2145
Specifications
Specification Test Results
- Identity:Determined by IR
Specification Test Results
About this item
:
:190.1 g/mol
Storage Temperature:Freezer
CAS Number:1198-77-2
Formula:C₅H₃N₄O₃·Na
MDL Number:MFCD00067310