Kaempferol
Glass bottle for solids, 50 mg
Catalog # ENZOALX385005M050
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Synonyms: 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
- CAS Number:520-18-3
- Formula:C₁₅H₁₀O₆
- MDL Number:MFCD00016938
- Molecular Weight:286.24 g/mol
- EINECS:208-287-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg



