Order Entry
Export
Orders LinkContactUsLinkComponent
 

Colchicine

Glass bottle for solids, 1 g
Catalog # ENZOALX380033G001
Toxic
Health
Supplier:   Enzo Life Sciences
Colchicine
Catalog # ENZOALX380033G001
Supplier:  Enzo Life Sciences
CAS Number:  64-86-8
Colchicine

Glass bottle for solids, 1 g

CAS Number:  64-86-8
Price
145,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  7α-H-Colchicine, (S)-(-)-Colchicine
  • CAS Number:64-86-8
  • Formula:C₂₂H₂₅NO₆
  • MDL Number:MFCD00078484
  • Molecular Weight:399.44 g/mol
  • Merck Index:12,02536
  • Melting Pt:154…156 °C
  • Density:1,32 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-598-5
  • UN:1544
  • ADR:6.1,I
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications