Order Entry
Export
Orders LinkContactUsLinkComponent
 

trans-Ned-19

Supplier:   Enzo Life Sciences
trans-Ned-19
Supplier:  Enzo Life Sciences
Certificates

2 Options Available Below

Product Details & Documents

Cell permeable, selective and potent inhibitor of nicotinic acid adenine dinucleotide phosphate (NAADP) receptor in mammalian cells and sea urchin eggs. Blocks NAADP signaling. Inhibits Ca2+ release: trans-Ned-19 (IC50 = 6nM); cis-Ned-19 (IC50 = 800nM). [32P] NAADP binding: trans-Ned-19 (IC50 = 0.4nM); cis-Ned-19 (IC50 = 15µM).
Inhibits Ca2+ release from lysosomes. Noncompetitive antagonist. Fluorescent.

Synthetic

  • CAS Number:1354235-96-3
  • Formula:C₃₀H₃₁FN₄O₃
  • MDL Number:MFCD02057367
  • Molecular Weight:514.601 g/mol
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 0,5 mg
    Log in for availability
    Log in for availability
    1 * 0,5 mg
    1 * 0,5 mg - 267,00 €
    267,00 €
    1 * 0,5 mg
       267,00 €

  • 1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 435,00 €
    435,00 €
    1 * 1 mg
       435,00 €