Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
CGP 74514A
CGP 74514A
CGP 74514A
CGP 74514A
ENZOALX270391M005
:
Enzo Life Sciences
:
CGP 74514A
ENZOALX270391M005
:
Enzo Life Sciences
:
Pk:
5 mg
Seq:
2210
:
:
385.9 g/mol
:
C₁₉H₂₄ClN₇