2,2-Bis(4-glycidyloxyphenyl)propane ≥85%
Catalog # ENZOALX270353G025
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:340.42 g/mol
CAS Number:1675-54-3
Formula:C₂₁H₂₄O₄
MDL Number:MFCD00080480