Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Piceatannol
Piceatannol
Piceatannol
Piceatannol
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
Piceatannol
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
10083-24-6
:
Formula:
C₁₄H₁₂O₄
MDL Number:
MFCD00221715
:
244.2427 g/mol