Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(-)-Glutathione (oxidized form)
L(-)-Glutathione (oxidised form)
L(-)-Glutathione (oxidised form)
L(-)-Glutathione (oxidised form)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
L(-)-Glutathione (oxidised form)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
27025-41-8
:
Formula:
C₂₀H₃₂N₆O₁₂S₂
MDL Number:
MFCD00063106
:
612.64 g/mol
Melting Pt:
185 °C
Storage Temperature:
Refrigerator
EINECS:
248-170-7