Orders LinkContactUsLinkComponent
(±)-10-Hydroxycamptothecin ≥97%
(±)-10-Hydroxycamptothecin ≥97%
(±)-10-Hydroxycamptothecin ≥97%
(±)-10-Hydroxycamptothecin ≥97%
A DNA topoisomerase I inhibitor that induces differentiation in HL-60 cells. It induces apoptosis in human breast cancer cell lines MCF-7 and MDA-MB-468.
  • Topoisomerase inhibitor
Pale yellow solid.
Soluble in DMSO.
 :  1-HCPT, 10-Hydroxycamptothecin, 4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, NSC107124
  •  :64439-81-2
  •  :C₂₀H₁₆N₂O₅
  •  :MFCD09027971
  •  :364.35 g/mol
  •  :Freezer
Product Family Options
  • Pk
  • 20 mg
    1 * 20 mg
    1 * 20 mg - 117,00 €
    117,00 €
    1 * 20 mg
       117,00 €
  • 100 mg
    1 * 100 mg
    1 * 100 mg - 439,00 €
    439,00 €
    1 * 100 mg
       439,00 €