Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
RG-13022
RG-13022 ≥98% (by TLC)
RG-13022 ≥98% (by TLC)
RG-13022 ≥98% (by TLC)
ENZOBMLEI2520005
:
ENZO LIFE SCIENCES
:
RG-13022 ≥98% (by TLC)
ENZOBMLEI2520005
:
ENZO LIFE SCIENCES
:
Pk:
5 mg
Seq:
2210
:
:
266.3 g/mol
:
Ambient
:
136831-48-6
:
C₁₆H₁₄N₂O₂
:
MFCD00236444