0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Benzylphosphonic acid-(AM)2
Benzylphosphonic acid-(AM)2 ≥98% (by TLC)
Benzylphosphonic acid-(AM)2 ≥98% (by TLC)
Benzylphosphonic acid-(AM)2 ≥98% (by TLC)
ENZOBMLEI2450010
:
ENZO LIFE SCIENCES
:
Benzylphosphonic acid-(AM)2 ≥98% (by TLC)
ENZOBMLEI2450010
:
ENZO LIFE SCIENCES
:
Pk:
10 mg
Seq:
2275
:
:
316.3 g/mol
:
38…39 °C
:
Freezer
:
200716-68-3
:
C₁₃H₁₇O₇P