Order Entry
Export
ContactUsLinkComponent
Rp-cAMPS ≥98% (by HPLC)
Rp-cAMPS ≥98% (by HPLC)
Catalog # ENZOBMLCN1350001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Rp-cAMPS ≥98% (by HPLC)
Catalog # ENZOBMLCN1350001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :446.5 g/mol
Storage Temperature:Freezer
CAS Number:151837-09-1
Formula:C₁₀H₁₁N₅O₅PS·C₆H₁₆N