Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
MMP-13 inhibitor
MMP-13 inhibitor ≥99% (by HPLC)
MMP-13 inhibitor ≥99% (by HPLC)
MMP-13 inhibitor ≥99% (by HPLC)
Supplier:
Enzo Life Sciences
Certificates
Expand image
MMP-13 inhibitor ≥99% (by HPLC)
Supplier:
Enzo Life Sciences
About this item
CAS Number:
544678-85-5
:
Formula:
C₂₂H₂₀F₂N₄O₂
MDL Number:
MFCD09878513
:
410.4 g/mol