Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
WYE-28
WYE-28 ≥97% (by HPLC)
WYE-28 ≥97% (by HPLC)
:
Enzo Life Sciences
WYE-28 ≥97% (by HPLC)
:
Enzo Life Sciences
:
1062172-60-4
:
:
586.6 g/mol