Order Entry
Export
ContactUsLinkComponent
Triacetyl resveratrol ≥98% (by TLC)
Corrosive
Irritant
Environment
Triacetyl resveratrol ≥98% (by TLC)
Catalog # ENZOBMLFR1190010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Corrosive
Irritant
Environment
Triacetyl resveratrol ≥98% (by TLC)
Catalog # ENZOBMLFR1190010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

 :
 :354.35 g/mol
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:42206-94-0
Formula:C₂₀H₁₈O₆
MDL Number:MFCD01546481