Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ZK-756326
ZK-756326 ≥98% (by TLC), white solid
ZK-756326 ≥98% (by TLC), white solid
:
Enzo Life Sciences
ZK-756326 ≥98% (by TLC), white solid
:
Enzo Life Sciences
:
874911-96-3
:
:
C₂₁H₂₈N₂O₃·2HCl
:
MFCD09038571
:
429.38 g/mol
Purity:
≥98% (TLC)