Order Entry
Export
ContactUsLinkComponent
Leukotriene C4 ≥95% (by HPLC)
Flammable
Toxic
Health
Leukotriene C4 ≥95% (by HPLC)
Catalog # ENZOBMLLC0040050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Flammable
Toxic
Health
Leukotriene C4 ≥95% (by HPLC)
Catalog # ENZOBMLLC0040050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Seq:
    1202

Specifications

Specification Test Results

  • Purity:
    ≥95% (HPLC)
  • Concentration:
    50 µg/ml
  • Source:
    Synthetic

Specification Test Results

About this item

 :
 :625.8 g/mol
Merck Index:14,05455
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
UN:1230
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:72025-60-6
Formula:C₃₀H₄₇N₃O₉S