Order Entry
Export
ContactUsLinkComponent
15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid ≥95% (by HPLC)
Flammable
15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid ≥95% (by HPLC)
Catalog # ENZOBMLK0150050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Flammable
15-Ketoeicosa-6E,8Z,11Z,13E-tetraenoic acid ≥95% (by HPLC)
Catalog # ENZOBMLK0150050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Seq:
    1202

Specifications

Specification Test Results

  • Purity:
    ≥95% (HPLC)
  • Concentration (for 50 µg size):
    50 µg/ml for 50 µg size
  • Concentration (for 1 mg size):
    1 mg/ml

Specification Test Results

About this item

 :
 :318.5 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
UN:1170
ADR:3,II
 :GHS02
CAS Number:81416-72-0