Order Entry
Export
Orders LinkContactUsLinkComponent
PD 98059 ≥95% (by HPLC)

PD 98059 ≥95% (by HPLC)

5 mg
Catalog # ENZOBMLEI3600005
Supplier:   Enzo Life Sciences
PD 98059 ≥95% (by HPLC)
Catalog # ENZOBMLEI3600005
Supplier:  Enzo Life Sciences
CAS Number:  167869-21-8
PD 98059 ≥95% (by HPLC)
PD 98059 ≥95% (by HPLC)

5 mg

CAS Number:  167869-21-8
Price
117,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

PD-98059 is a potent and selective inhibitor of MAP kinase kinase (MEK). It selectively blocks the activation of MEK thereby inhibiting the phosphorylation and the activation of MAP kinase. PD-98059 is cell permeable. In PC12 pheochromocytoma cells, it completely blocked the increase in MAP kinase activity produced by NGF, IC50=2µM. Enhances self-renewal of embryonic stem cells PD-98059 is an invaluable tool to help elucidate the role of the MAPK cascade in a variety of biological systems. Inhibits cell growth and proliferation in acute myelogenous leukemia cell lines by causing cell arrest.

  • MEK inhibitor

Pale yellow solid

Soluble in DMSO (6.5mg/ml); 25mg/ml in anhydrous DMSO), methanol, dichlormethane and 100% ethanol (0.6mg/ml).

Synonyms:  PD98059, 2'-Amino-3'-methoxyflavone, 2-(2-Amino-3-methoxyphenyl)chromone, PD 98059, 2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, 2-(2-Amino-3-methoxyphenyl)-4H-chromen-4-one
  • CAS Number:167869-21-8
  • Formula:C₁₆H₁₃NO₃
  • MDL Number:MFCD00671789
  • Molecular Weight:267.284 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pk:
    5 mg

Specifications