Order Entry
Export
ContactUsLinkComponent
4,4-Pentamethylene-2-pyrrolidone ≥95%
4,4-Pentamethylene-2-pyrrolidone ≥95%
Catalog # ENZOBMLKC1600250
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
4,4-Pentamethylene-2-pyrrolidone ≥95%
Catalog # ENZOBMLKC1600250
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :153.23 g/mol
CAS Number:64744-50-9
Formula:C₉H₁₅NO
MDL Number:MFCD00177938