Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acridine orange (chlorid salt)
Acridine orange (chlorid salt) ≥95% (by HPLC), Ultrapure
Acridine orange (chlorid salt) ≥95% (by HPLC), Ultrapure
Acridine orange (chlorid salt) ≥95% (by HPLC), Ultrapure
ENZOENZ52405
:
ENZO LIFE SCIENCES
:
Acridine orange (chlorid salt) ≥95% (by HPLC), Ultrapure
ENZOENZ52405
:
ENZO LIFE SCIENCES
:
Pk:
100 mg
Seq:
2440
:
:
301.81 g/mol (anhy)
:
Freezer
:
65-61-2
:
1704465-79-1
:
C₁₇H₁₉N₃·HCl·xH₂O
:
MFCD00012660