Order Entry
Export
ContactUsLinkComponent
17-Octadecynoic acid ≥99%
17-Octadecynoic acid ≥99%
Catalog # ENZOBMLEI1030010
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
17-Octadecynoic acid ≥99%
Catalog # ENZOBMLEI1030010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :280.45 g/mol
CAS Number:34450-18-5
Formula:C₁₈H₃₂O₂