Order Entry
Export
ContactUsLinkComponent
3-Methylcyclopentadecanone ≥99% (by TLC), colourless oil
3-Methylcyclopentadecanone ≥99% (by TLC), colourless oil
Catalog # ENZOBMLEI1610500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
3-Methylcyclopentadecanone ≥99% (by TLC), colourless oil
Catalog # ENZOBMLEI1610500
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :238.41 g/mol
Storage Temperature:Ambient
CAS Number:541-91-3
Formula:C₁₆H₃₀O
MDL Number:MFCD00211114