ContactUsLinkComponent
Oakton® Conductivity and TDS Standards
  VWRU76338-556
 :  
undefined
Oakton® Conductivity and TDS Standards
  VWRU76338-556
 :  
  • Pk:
    500 mL
  • Description:
    Conductivity and TDS standard
  • TDS calibration:
    225,6 (ppm KCl)
    215,5 (ppm NaCl)
  • TDS conversion factor:
    300 (442 ppm)
  • Conductivity:
    447 µS/cm
Ideal for laboratory and field analysis.
  • Traceable to NIST standard reference materials
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25 °C
Oakton conducitivity and TDS standards feature temperature corrections, KCl, NaCL, and 442 conversions printed directly on the bottle for ease of use (excluded on 60-mL bottle, but included in insert).
 : These standards are manufactured under ISO 9000 quality guidelines and traceable to NIST standards.
 : If unopened, shelf life is one year. If opened, shelf life if 4 months.